siRNA Details
Gene Symbol: Tssk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007650.1 | NP_001007651.1 | 1 | 194 | ||
GenBank mRNA | AY346102.1 | - | 1 | 194 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_080442, AF298901, AK005983, AK019840, BC048470, and others.
Gene Ontology Categories:
