siRNA Details
Gene Symbol: Pef1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007651.1 | NP_001007652.1 | 4 | 578 | ||
GenBank mRNA | BC082028.1 | - | 4 | 578 | 7130077 | |
FQ211313.1 | - | 4 | 575 | |||
FQ211450.1 | - | 4 | 591 | |||
FQ219120.1 | - | 4 | 591 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
