siRNA Details
Gene Symbol: Wwp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024757.1 | NP_001019928.1 | 11 | 1490 | ||
XM_006237930.2 | XP_006237992.1 | 11 | 1405 | |||
XM_006237931.2 | XP_006237993.1 | 10 | 1832 | |||
XM_008763590.1 | XP_008761812.1 | 12 | 1703 | |||
XM_008763591.1 | XP_008761813.1 | 4 | 898 | |||
GenBank mRNA | BC097386.1 | - | 11 | 1490 | 7446512 |
