siRNA Details
Gene Symbol: Smim8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106644.1 | NP_001100114.1 | 4 | 722 | ||
NM_001201373.1 | NP_001188302.1 | 2 | 198 | |||
NM_001201374.1 | NP_001188303.1 | 3 | 344 | |||
NM_001201375.1 | NP_001188304.1 | 3 | 588 | |||
XM_006237987.2 | XP_006238049.1 | 4 | 627 | |||
XM_006237988.2 | XP_006238050.1 | 3 | 510 | |||
GenBank mRNA | BC167112.1 | - | 2 | 198 | 6887495 | |
DY544415.1 | - | 3 | 344 | |||
FQ226097.1 | - | 3 | 549 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
