siRNA Details
Gene Symbol: Chmp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025410.1 | NP_001020581.1 | 4, 5 | 363 | ||
GenBank mRNA | BC086333.1 | - | 3, 4 | 225 | ||
BC097963.1 | - | 4, 5 | 363 | 7387550 | ||
FQ214132.1 | - | 4, 5 | 391 | |||
FQ228147.1 | - | 4, 5 | 397 | |||
FQ234681.1 | - | 4, 5 | 397 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
