siRNA Details
Gene Symbol: Rad23b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025275.1 | NP_001020446.1 | 3 | 472 | ||
GenBank mRNA | BC098674.1 | - | 3 | 472 | 7190031 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_009011, BC068193, BC027747, AK160890, AK157252, and others.
Gene Ontology Categories:
