siRNA Details
Gene Symbol: Rad23b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025275.1 | NP_001020446.1 | 8 | 1156 | ||
GenBank mRNA | BC070960.1 | - | 4 | 375 | ||
BC090351.1 | - | 5 | 619 | |||
BC098674.1 | - | 8 | 1156 | 7190031 | ||
BC111406.1 | - | 3 | 295 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_009011, AK193972, AK198214, BC068193, BC027747, and others.
Gene Ontology Categories:
