siRNA Details
Gene Symbol: Slc46a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106652.1 | NP_001100122.1 | 3 | 1418 | ||
XM_008763734.1 | XP_008761956.1 | 2 | 1592 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Slc46a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106652.1 | NP_001100122.1 | 3 | 1418 | ||
XM_008763734.1 | XP_008761956.1 | 2 | 1592 |