siRNA Details
Gene Symbol: Slc46a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106652.1 | NP_001100122.1 | 1 | 506 | ||
XM_008763734.1 | XP_008761956.1 | 1 | 764 |

More Information
Other Species Targeted

Gene Ontology Categories:
