siRNA Details
Gene Symbol: Erp44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008317.1 | NP_001008318.1 | 4 | 275 | ||
GenBank mRNA | AY158662.1 | - | 4 | 275 | ||
FQ211910.1 | - | 369 | ||||
FQ227188.1 | - | 4 | 335 | |||
FQ234126.1 | - | 4 | 336 | |||
FQ234429.1 | - | 4 | 376 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_029572, AK172973, BC019558, AK160113, AK003217, and others.
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
