siRNA Details
Gene Symbol: Sec61b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106654.2 | NP_001100124.1 | 4 | 446 | ||
GenBank mRNA | BC167110.1 | - | 4 | 446 | 6887572 | |
FQ210322.1 | - | 4 | 434 | |||
FQ217838.1 | - | 4 | 399 | |||
FQ221073.1 | - | 4 | 393 | |||
FQ221908.1 | - | 4 | 390 |

More Information
Other Species Targeted

Gene Ontology Categories:
