siRNA Details
Gene Symbol: RGD1305807 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025276.1 | NP_001020447.1 | 7, 8 | 651 | ||
XM_006238039.1 | XP_006238101.1 | 6, 7 | 568 | |||
GenBank mRNA | BC097368.1 | - | 7, 8 | 651 | 7443399 |
