siRNA Details
Gene Symbol: Zbtb5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106657.1 | NP_001100127.1 | 2 | 938 | ||
XM_006238046.2 | XP_006238108.1 | 2 | 936 | |||
XM_006238047.2 | XP_006238109.1 | 2 | 1569 | |||
XM_006238048.2 | XP_006238110.1 | 2 | 2219 |

More Information
Other Species Targeted

Gene Ontology Categories:
