siRNA Details
Gene Symbol: Zbtb5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106657.1 | NP_001100127.1 | 2 | 925 | ||
XM_006238046.2 | XP_006238108.1 | 2 | 923 | |||
XM_006238047.2 | XP_006238109.1 | 2 | 1556 | |||
XM_006238048.2 | XP_006238110.1 | 2 | 2206 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001163284, NM_173399, NM_001163283, AK031172, AK160003, and others.
Gene Ontology Categories:
