siRNA Details
Gene Symbol: Slc31a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033693.2 | NP_001028865.2 | 3 | 323 | ||
XM_006238230.2 | XP_006238292.1 | 2 | 291 | |||
GenBank mRNA | BC099751.1 | - | 3 | 296 | 5623519 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
