siRNA Details
Gene Symbol: LOC298139 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013930.1 | NP_001013952.1 | 2 | 453 | ||
XM_006238294.1 | XP_006238356.1 | 2 | 508 | |||
GenBank mRNA | BC083698.1 | - | 2 | 453 | 7189881 | |
FQ217875.1 | - | 2 | 508 |
