siRNA Details
Gene Symbol: Zdhhc21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039009.1 | NP_001034098.1 | 7 | 848 | ||
XM_008763765.1 | XP_008761987.1 | 7 | 3750 | |||
XM_008763766.1 | XP_008761988.1 | 8 | 1046 | |||
XM_008763767.1 | XP_008761989.1 | 8 | 905 | |||
XM_008763768.1 | XP_008761990.1 | 8 | 3459 | |||
GenBank mRNA | AY886536.1 | - | 7 | 848 |
