siRNA Details
Gene Symbol: Frem1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191757.1 | NP_001178686.1 | 18 | 3552 | ||
XM_006238349.2 | XP_006238411.1 | 19 | 4167 | |||
XM_008763769.1 | XP_008761991.1 | 19 | 4168 | |||
XM_008763770.1 | XP_008761992.1 | 18 | 3653 | |||
XM_008763771.1 | XP_008761993.1 | 15 | 2630 |
