siRNA Details
Gene Symbol: Dhcr24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080148.1 | NP_001073617.1 | 7 | 1123 | ||
GenBank mRNA | AY310319.1 | - | 7 | 1123 | ||
AY921220.2 | - | 7 | 1185 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Dhcr24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080148.1 | NP_001073617.1 | 7 | 1123 | ||
GenBank mRNA | AY310319.1 | - | 7 | 1123 | ||
AY921220.2 | - | 7 | 1185 |