siRNA Details
Gene Symbol: Nkrf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_233308.8 | XP_233308.7 | 1585 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Nkrf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_233308.8 | XP_233308.7 | 1585 |