siRNA Details
Gene Symbol: Ube2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013933.1 | NP_001013955.1 | 6 | 458 | ||
XM_006257446.2 | XP_006257508.1 | 5 | 471 | |||
GenBank mRNA | BC079353.1 | - | 6 | 458 | 7108236 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_019668, AK164074, BC026053, AK150417, AF089812, and others.
Gene Ontology Categories:
