siRNA Details
Gene Symbol: Rhox9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024874.1 | NP_001020045.1 | 2 | 102 | ||
GenBank mRNA | DQ058656.1 | - | 2 | 102 |
Gene Symbol: LOC100364002 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006257483.2 | XP_006257545.1 | 2 | 94 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
