siRNA Details
Gene Symbol: Elavl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006238591.2 | XP_006238653.1 | 7 | 1672 | ||
XM_006238592.2 | XP_006238654.1 | 7 | 1987 | |||
XM_006238593.2 | XP_006238655.1 | 7 | 1420 | |||
XM_006238594.2 | XP_006238656.1 | 7 | 1696 | |||
XM_006238595.2 | XP_006238657.1 | 7 | 1629 | |||
XM_008763893.1 | XP_008762115.1 | 7 | 1556 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001163399, NM_001038698, NM_001163397, NM_010488, AK079088, and others.
Gene Ontology Categories:
