siRNA Details
Gene Symbol: Elavl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006238591.2 | XP_006238653.1 | 7 | 2236 | ||
XM_006238592.2 | XP_006238654.1 | 7 | 2551 | |||
XM_006238593.2 | XP_006238655.1 | 7 | 1984 | |||
XM_006238594.2 | XP_006238656.1 | 7 | 2260 | |||
XM_006238595.2 | XP_006238657.1 | 7 | 2193 | |||
XM_008763893.1 | XP_008762115.1 | 7 | 2120 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
