siRNA Details
Gene Symbol: Coa7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106674.1 | NP_001100144.1 | 3 | 556 |

More Information
Other Species Targeted


UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Coa7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106674.1 | NP_001100144.1 | 3 | 556 |