siRNA Details
Gene Symbol: RGD1559786 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034132.1 | NP_001029304.1 | 2 | 113 | ||
XM_006238508.2 | XP_006238570.1 | 2 | 178 | |||
GenBank mRNA | BC100100.1 | - | 2 | 113 | 7376782 | |
FQ228722.1 | - | 2 | 132 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
