siRNA Details
Gene Symbol: Ccin | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005904.1 | NP_001005904.1 | 1 | 1231 | ||
GenBank mRNA | BC083832.1 | - | 1 | 1231 | 7112736 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ccin | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005904.1 | NP_001005904.1 | 1 | 1231 | ||
GenBank mRNA | BC083832.1 | - | 1 | 1231 | 7112736 |