siRNA Details
Gene Symbol: Ccin | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005904.1 | NP_001005904.1 | 1 | 817 | ||
GenBank mRNA | BC083832.1 | - | 1 | 817 | 7112736 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
