siRNA Details
Gene Symbol: Fam221b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013934.1 | NP_001013956.1 | 6 | 1533 | ||
XM_008763669.1 | XP_008761891.1 | 6 | 1758 | |||
XM_008763670.1 | XP_008761892.1 | 6 | 1743 | |||
GenBank mRNA | BC081906.1 | - | 6 | 1533 | 7111080 |
