siRNA Details
Gene Symbol: Creb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013092.2 | NP_001013110.1 | 4 | 580 | ||
XM_006238050.1 | XP_006238112.1 | 4 | 606 | |||
XM_008763672.1 | XP_008761894.1 | 5 | 410 | |||
GenBank mRNA | BC062241.1 | - | 4 | 583 | 6887761 | |
KJ817427.1 | - | 4 | 424 |
