siRNA Details
Gene Symbol: Ipp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106679.2 | NP_001100149.2 | 8 | 1705 | ||
XM_006238640.2 | XP_006238702.1 | 9 | 1877 | |||
XM_006238642.2 | XP_006238704.1 | 9 | 1888 | |||
XM_006238645.2 | XP_006238707.1 | 8 | 1751 | |||
XM_008763992.1 | XP_008762214.1 | 9 | 1888 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
