siRNA Details
Gene Symbol: Dph2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001015007.1 | NP_001015007.1 | 4 | 997 | ||
GenBank mRNA | BC092656.1 | - | 4 | 997 | 7320904 |

More Information
Other Species Targeted

Gene Ontology Categories:
