siRNA Details
Gene Symbol: Ppcs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039010.2 | NP_001034099.2 | 1 | 443 | ||
GenBank mRNA | BC099208.1 | - | 1 | 833 | 7384489 | |
CK474267.1 | - | 1 | 482 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
