siRNA Details
Gene Symbol: Mrps15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007653.1 | NP_001007654.1 | 5 | 407 | ||
GenBank mRNA | BC083856.1 | - | 5 | 407 | 7125450 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
