siRNA Details
Gene Symbol: Csf3r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106685.1 | NP_001100155.1 | 12 | 1722 | ||
XM_008764003.1 | XP_008762225.1 | 12 | 1993 | |||
XM_008764004.1 | XP_008762226.1 | 11 | 1902 | |||
XM_008764005.1 | XP_008762227.1 | 12 | 1821 | |||
XM_008764006.1 | XP_008762228.1 | 11 | 1730 | |||
XM_008764007.1 | XP_008762229.1 | 12 | 1960 |
