siRNA Details
Gene Symbol: Dhdds | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011978.1 | NP_001011978.1 | 4 | 441 | ||
XM_006239014.2 | XP_006239076.1 | 4 | 556 | |||
GenBank mRNA | BC085875.1 | - | 4 | 441 | 7191470 |

More Information
Other Species Targeted

Gene Ontology Categories:
