siRNA Details
Gene Symbol: Sdhb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100539.1 | NP_001094009.1 | 2 | 200 | ||
GenBank mRNA | BC158620.1 | - | 2 | 200 | 7446362 | |
DQ403001.1 | - | 1 | 105 | |||
FQ215046.1 | - | 2 | 196 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
