siRNA Details
Gene Symbol: Nmnat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037556.2 | NP_001032645.2 | 5 | 866 | ||
XM_006239429.2 | XP_006239491.1 | 5 | 1500 | |||
XM_006239430.2 | XP_006239492.1 | 5 | 1085 | |||
XM_006239431.2 | XP_006239493.1 | 4 | 827 | |||
XM_008764237.1 | XP_008762459.1 | 6 | 1023 | |||
XM_008764238.1 | XP_008762460.1 | 5 | 848 | |||
GenBank mRNA | AY788917.1 | - | 5 | 860 | ||
BC127446.1 | - | 5 | 866 | 7383226 |
