siRNA Details
Gene Symbol: H6pd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106698.1 | NP_001100168.1 | 5 | 1398 | ||
XM_006239459.2 | XP_006239521.1 | 5 | 1548 | |||
XM_006239460.2 | XP_006239522.1 | 6 | 1683 |
