siRNA Details
Gene Symbol: Rer1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039012.1 | NP_001034101.1 | 4 | 266 | ||
GenBank mRNA | BC100270.1 | - | 4 | 266 | 7444634 |

More Information
Other Species Targeted

Gene Ontology Categories:
