siRNA Details
Gene Symbol: Vwa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013938.1 | NP_001013960.1 | 2 | 306 | ||
GenBank mRNA | BC081983.1 | - | 2 | 306 | 7128587 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
