siRNA Details
Gene Symbol: Aurkaip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004237.1 | NP_001004237.1 | 3 | 513 | ||
XM_006239546.2 | XP_006239608.1 | 3 | 701 | |||
XM_006239547.2 | XP_006239609.1 | 3 | 911 | |||
XM_006239548.2 | XP_006239610.1 | 3 | 658 | |||
XM_008764351.1 | XP_008762573.1 | 3 | 534 | |||
GenBank mRNA | BC079171.1 | - | 3 | 513 | 7129954 | |
FQ215450.1 | - | 3 | 921 | |||
FQ228270.1 | - | 3 | 521 | |||
FQ232591.1 | - | 3 | 521 |

More Information
Other Species Targeted

Gene Ontology Categories:
