siRNA Details
Gene Symbol: Gpalpp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024875.1 | NP_001020046.1 | 5, 6 | 648 | ||
GenBank mRNA | BC097485.1 | - | 5, 6 | 648 | 7459542 | |
FQ214079.1 | - | 336 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
