siRNA Details
Gene Symbol: Crim1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001169103.1 | NP_001162574.1 | 7, 8 | 1901 | ||
XM_006239623.2 | XP_006239685.1 | 7, 8 | 1958 | |||
XM_006239625.2 | XP_006239687.1 | 6, 7 | 1715 |
