siRNA Details
Gene Symbol: Crim1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001169103.1 | NP_001162574.1 | 16 | 3293 | ||
XM_006239623.2 | XP_006239685.1 | 16 | 3347 | |||
XM_006239625.2 | XP_006239687.1 | 15 | 3107 | |||
GenBank mRNA | FQ217152.1 | - | 14 | 2093 |
