siRNA Details
Gene Symbol: Zfp36l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001036626.1 | NP_001031703.1 | 2 | 571 | ||
XM_008764455.1 | XP_008762677.1 | 2 | 2635 | |||
GenBank mRNA | CV112816.1 | - | 2 | 571 |
Gene Symbol: LOC100911319 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750261.1 | XP_003750309.1 | 2 | 571 | ||
XM_008765049.1 | XP_008763271.1 | 2 | 600 |
