siRNA Details
Gene Symbol: Prepl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010951.1 | NP_001010951.1 | 8 | 1219 | ||
XM_006239697.2 | XP_006239759.1 | 9 | 1782 | |||
XM_006239698.2 | XP_006239760.2 | 9 | 1347 | |||
XM_006239699.2 | XP_006239761.2 | 9 | 2059 | |||
XM_008764456.1 | XP_008762678.1 | 9 | 1449 | |||
XM_008764457.1 | XP_008762679.1 | 9 | 1342 | |||
XM_008764458.1 | XP_008762680.1 | 8 | 1142 | |||
GenBank mRNA | BC089111.1 | - | 8 | 1219 | 7312962 | |
FQ213992.1 | - | 2 | 258 |
