siRNA Details
Gene Symbol: Memo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029917.1 | NP_001025088.1 | 6, 7 | 691 | ||
GenBank mRNA | BC098061.1 | - | 6, 7 | 691 | 7458219 | |
FQ215386.1 | - | 4, 5 | 443 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_133771, AF363447, AK002639, AK159560, BC013819.
Gene Ontology Categories:
