siRNA Details
Gene Symbol: Emilin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006239793.2 | XP_006239855.1 | 2, 3 | 774 | ||
XM_008764511.1 | XP_008762733.1 | 2, 3 | 1506 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Emilin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006239793.2 | XP_006239855.1 | 2, 3 | 774 | ||
XM_008764511.1 | XP_008762733.1 | 2, 3 | 1506 |