siRNA Details
Gene Symbol: Mapre3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007656.1 | NP_001007657.1 | 7 | 744 | ||
XM_006239796.2 | XP_006239858.1 | 6 | 627 | |||
XM_008764512.1 | XP_008762734.1 | 6 | 794 | |||
XM_008764513.1 | XP_008762735.1 | 6 | 749 | |||
GenBank mRNA | BC083589.1 | - | 7 | 744 | 7132546 |

More Information
Other Species Targeted

Gene Ontology Categories:
